Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine dihydrochloride, 99%

Catalog No. AC411760025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411760025 2.5 kg Plastic Bottle
AC411761000 100 g Glass bottle
AC411765000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC411760025 Supplier Thermo Scientific Chemicals Supplier No. 411760025
Add to Cart

CAS: 5341-61-7 | ClH5N2 | 68.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5341-61-7
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
Synonym hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
PubChem CID 17548
IUPAC Name hydrazine;dihydrochloride
SMILES [H+].[Cl-].NN

Specifications

Melting Point 198.0°C
Color White
Physical Form Chunks and/or Powder
Quantity 2.5 kg
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula NH2NH2·2HCl
Merck Index 15, 4809
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
Formula Weight 104.97
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Plastic Bottle
Chemical Name or Material Hydrazine dihydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Gently
EINECSNumber 226-283-2
RTECSNumber MV2298000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.