Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine Hydrate, Technical, 85%, Spectrum™ Chemical
SDP

Catalog No. 18610829 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
10 mL
250 mL
1 L
5 gal.
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Drum

Catalog No. 18-610-829

Supplier: Spectrum Chemical Manufacturing Corporation H10141LT

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7803-57-8

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 L
Percent Purity 0.85
Grade Technical

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.