Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine monohydrochloride, 98+%

Catalog No. p-3778763
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum

CAS: 2644-70-4 | H4N2·HCl | 68.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2644-70-4
Molecular Formula H4N2·HCl
Molecular Weight (g/mol) 68.51
MDL Number MFCD00044368
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
Synonym hydrazine hydrochloride, hydrazine monohydrochloride, hydrazine, monohydrochloride, hydrazine,chloride, diamine hydrochloride, hydrazine.hydrochloride, hydrazine,hydrochloride, hydrazine mono hcl, nh2nh2hcl, pubchem17422
PubChem CID 115031
IUPAC Name hydrazine;hydrochloride
SMILES NN.Cl

Specifications

Melting Point 89°C to 93°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 98% min. (Argentometry)
Linear Formula H2NNH2·HCl
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: slightly soluble in alcohol,insoluble in most organic solvents,soluble in liquid ammoniak
Formula Weight 68.51
Percent Purity ≥98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Hydrazine monohydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
EINECSNumber 220-154-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.