Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine Standard, 100 ppm N2H4, Ricca Chemical
SDP

Catalog No. 355216 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3552-16 500 mL Amber Glass Bottle
3552-4 120 mL Amber Glass Bottle
3552-32 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 3552-16 Supplier Ricca Chemical Company Supplier No. 355216
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5341-61-7 , 7647-01-0 , 7732-18-5
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
PubChem CID 17548
IUPAC Name hydrogen hydrazine chloride
SMILES [H+].[Cl-].NN

Specifications

Packaging Amber Glass Bottle
CAS Min % 97.0, 1.14, 0.03
CAS Max % 100.0, 1.19, 0.03
Equivalence 1 mL = 0.1 mg N
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Hydrazine Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.