Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine sulfate, 99+%

Catalog No. aaa1079222
Click to view available options
Quantity:
100 g
250 g
1 kg

CAS: 10034-93-2 | H6N2O4S | 130.118 g/mol

Hydrazine sulfate is used in analytical chemistry for the gravimetric estimation of nickel, cobalt and cadmium. It is also used in the refining of rare metals as well as in the separation of polonium from tellurium. It finds application in the synthesis of chemical intermediate and organic compounds. Further, it acts as an antioxidant in soldering flux for light metals and as a precursor to hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrazine sulfate is used in analytical chemistry for the gravimetric estimation of nickel, cobalt and cadmium. It is also used in the refining of rare metals as well as in the separation of polonium from tellurium. It finds application in the synthesis of chemical intermediate and organic compounds. Further, it acts as an antioxidant in soldering flux for light metals and as a precursor to hydrazine.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.comIncompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-93-2
Molecular Formula H6N2O4S
Molecular Weight (g/mol) 130.118
MDL Number MFCD00044873
InChI Key ZGCHATBSUIJLRL-UHFFFAOYSA-N
Synonym hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate
PubChem CID 24842
IUPAC Name hydrazine;sulfuric acid
SMILES NN.OS(=O)(=O)O

Specifications

Melting Point 254°C
Density 1.37
pH 1.3
Odor Odorless
Quantity 100 g
UN Number UN3288
Merck Index 14,4772
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 130.12
Percent Purity ≥99%
Chemical Name or Material Hydrazine sulfate
Show More Show Less
Hazard Category H301+H311+H331-H317-H350
Hazard Statement GHS H Statement
H311-H331-H350-H302-H317
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 233-110-4
RTECSNumber MV9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.