Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrazobenzene 98.0+%, TCI America™

Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
Melting Point | 127°C |
Color | White-Yellow |
Quantity | 100 g |
UN Number | 3077 |
Formula Weight | 184.24 |
Percent Purity | ≥98.0% (HPLC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Hydrazobenzene |
Safety and Handling
EINECSNumber | (3)-2755 |
RTECSNumber | MW2625000 |
TSCA | Yes |
Recommended Storage | Refrigerator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction