Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Micro Essential Lab Hydrion™ Buffer Chemvelopes
Shop All Micro Essential Lab Products
Click to view available options
pH:
2
3
4
6.4
7
8
9
10
11
12
Physical Form:
Capsule
Chemvelope
Description
- In capsules or packets (Chemvelopes)
- Dissolve in distilled water: 100mL for capsules, 500mL for packets
- Directly traceble to NIST standards
Standard Capsules come in vials of 10, five vials per pack. Tri-Chek Capsule Set includes one vial of 10 capsules each for pH 4, 7, and 10 buffers
Tri-Chek Chemvelope Set includes three pouches each for pH 4 and 10, four pouches for pH 7
Tri-Chek sets also include dispensing dropper bottle of Color Key/Buffer Preservative; add to buffer to inhibit mold and to color-code solution: red-orange for pH 4, green for pH 7, blue for pH 10
Specifications
Specifications
| pH | 4 |
| Physical Form | Capsule |
| Quantity | Each capsule makes 100 mL solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction