Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrion™ pH Test Papers, Wide-Range
Shop All Micro Essential Lab Products

Click to view available options
pH Range:
1 to 11
2 to 10
Type:
Hydrion D/R AB Dispenser
Refill Roll
Type A Refill
Type A Vial
Type B pH Paper
Quantity:
10 Pack
5 Pack
Each
Description
- Type A indicates pH 2, 4, 6, 8 and 10
- Type B indicates pH 1, 3, 5, 7, 9 and 11
Order Info
- Cat. No. S60146 and S60148: 5 rolls and 2 color charts per kit, 10 kits per case
- Cat. No. S60152: 10 vials per box
Specifications
Specifications
| pH Range | 1 to 11 |
| For Use With (Application) | To determine pH within half unit via distinct color changes |
| Includes | Color comparison scale |
| Quantity | 5 Pack |
| Type | Type A Refill |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction