Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic Acid, 48%, ACS, 47.0-49.0%, Spectrum™ Chemical
SDP

Catalog No. 18610842 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-842 200 L Drum
18-610-841 120 mL Amber Glass Bottle
18-610-849 500 mL Amber Glass Bottle
18-610-848 2.5 L Poly-Coated Glass Bottle
18-610-840 4 L Amber Glass Bottle
18-610-843 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-610-842 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H1030200LTBL
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5

Specifications

Concentration 48%
CAS Max % 40 to 60%
Heavy Metals 5 ppm
Packaging Drum
Residue after Ignition 2e-05
Quantity 200 L
Percent Purity 47 to 49%
Grade ACS
Chemical Name or Material Hydrobromic Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.