Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic Acid, 48%, Technical, Spectrum™ Chemical
SDP

Catalog No. 18610844 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
200 L
Packaging:
Poly-Coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-844 2.5 L Poly-Coated Glass Bottle
18-610-847 500 mL Poly-Coated Glass Bottle
18-610-846 4 L Poly-Coated Glass Bottle
18-610-845 200 L Poly-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-844 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H10312.5LTPV
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5

Specifications

CAS Max % 40 to 60%
Packaging Poly-Coated Glass Bottle
Quantity 2.5 L
Grade Technical
Chemical Name or Material Hydrobromic Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.