Hydrobromic Acid, ACS, 47.0-49%, Alfa Aesar

Air Sensitive

Manufacturer: Alfa Aesar 36694K2

Quantity 1L

 View more versions of this product

Catalog No. AA36694K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10035-10-6,7732-18-5
    Physical Form Liquid
    Assay Percent Range 47.0-49.0%
    Molecular Formula HBr
    Formula Weight 80.92
    MDL Number MFCD00011323
    Synonym Hydrogen bromide
    Chemical Name or Material Hydrobromic acid
    Grade ACS Grade

    Specifications

    Physical Form Liquid
    Formula Weight 80.92
    Quantity 1L
    Color Colorless to pale yellow
    DOT Information Hazard Class: 8, Packing Group: II
    EINECS Number 233-113-0
    CAS 10035-10-6,7732-18-5
    Concentration or Composition (by Analyte or Components) Hydrogen bromide: 48.0%; Water: 52.0%
    Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
    Vapor Pressure 22500 hPa (16876mm Hg) at 20°C
    Assay Percent Range 47.0-49.0%
    Molecular Formula HBr
    MDL Number MFCD00011323
    Synonym Hydrogen bromide
    Appearance Colorless to pale yellow liquid
    Chemical Name or Material Hydrobromic acid
    Grade ACS Grade

    H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit