Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic acid, pure, ca. 48 wt% solution in water

Catalog No. p-21396
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle

CAS: 10035-10-6 | BrH | 80.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6, 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name bromane
SMILES Br

Specifications

Color Brown to Yellow
Melting Point -11.0°C
Boiling Point 126.0°C to 128.0°C
Density 1.4800g/mL
Packaging Glass bottle
Specific Gravity 1.48
Quantity 1 L
Physical Form Solution
Linear Formula HBr
Fieser 01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273
Merck Index 15, 4816
Solubility Information Solubility in water: soluble.
Formula Weight 80.9
Percent Purity ≥47%
Grade Pure
Chemical Name or Material Hydrobromic acid, tech.
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 233-113-0
RTECSNumber MW3850000
TSCA TSCA
Recommended Storage Slowly darkens on exposure to air and light

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.