Learn More
3B Scientific™ Hydrocar
SDPSCI_ED
Catalog No. s96258 Shop All 3B Scientific Products
Change view
Click to view available options
:
Hydrocar
Catalog No.
S96258 Hydrocar
1 options
Catalog No. S96258
Supplier: 3B Scientific™ U40622
Only null left
Add to Cart
Add to Cart
New design offers futuristic feel for what transportation could look like in the next decades
This vehicle comes straight from the year 2099 and runs on clean hydrogen fuel using a next generation reversible Polymer Electrolyte Membrane (PEM) fuel cell. It combines water electrolysis and fuel cell functions into one device.
  • Add water to tanks in back of car
  • Watch as oxygen and hydrogen gases form in two transparent containers
  • Turns around obstacles by itself
  • Steers independently of the user once it hits a barrier
  • Blue LED lights flash from inside the cockpit
Specifications
Product Type Hydrocar