Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrocarbon Mixture No. 1 for ASTM D896, Reagents

Catalog No. 01334977
Change view
Click to view available options
Quantity:
950 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-977 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-977 Supplier Reagents Holdings Llc Supplier No. CH180000950C
Only null left
Add to Cart
Add to Cart

Hydrocarbon Mixture No. 1 for ASTM D896, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 540-84-1 , 108-88-3
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Packaging Glass Bottle
Specific Gravity 0.7567
Physical Form Liquid
Quantity 950 mL
Grade Reagent
Chemical Name or Material Hydrocarbon Mixture No. 1 for ASTM D896
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.