Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ricca Chemical Company Hydrochloric Acid, 0.0050 Normal, from NF/FCC Materials
SDP

Catalog No. R35890001A Shop All Ricca Chemical Company Products
Click to view available options
:
Hydrochloric Acid; Quantity: 1 L; Packaging: Poly

Hydrochloric Acid is tested with the tightest specifications in the industry for low lot to lot variability.

  • Manufactured in cGMP compliant, FDA registered facilities.
  • Certified Traceable to NIST Standard Reference Material when available.
  • Many C of As and labels contain TDS and conductance information.
  • Tightest Specifications in the industry, lowest lot-to-lot variability.

Specifications

Chemical Name or Material Hydrochloric Acid
Appearance of Solution Colorless
CAS 7732-18-5, 7647-01-0, 7647-01-0, 7732-18-5
Concentration 0.00500 (Normal at 20°C)
CAS Min % 0.02%, 99.98%
Packaging Poly
CAS Max % 99.98%, 0.02%
Traceability to NIST SRM# 723
Certificate of Analysis Yes
Assay Percent Range 0.00495 to 0.00505%
Assay Percent Notes Normal at 20°C
Quantity 1 L
Physical Form Liquid
Molecular Weight (g/mol) 36.46 g/mol
Grade Laboratory
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.