Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.0100 N (N/100), Ricca Chemical
SDP

Catalog No. 35905 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5, 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Appearance of Solution Colorless Liquid
Concentration 0.00995 to 0.01005 N
CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Packaging Cubitainer
Quantity 20 L
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.0100 N
Assay Normal at 20°C
Chemical Name or Material Hydrochloric Acid
Show More Show Less
ShelfLife 24 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.