Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.1 Normal (N/10) in Isopropanol with Bromophenol Blue, Reagents

Catalog No. 01335084
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-084 100 mL
01-335-083 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-084 Supplier Reagents Holdings Llc Supplier No. CH103800100C
Only null left
Add to Cart
Add to Cart

Hydrochloric Acid, 0.1 Normal (N/10) in Isopropanol with Bromophenol Blue, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Hydrochloric Acid, 0.1 Normal (N/10) in Isopropanol with Bromophenol Blue
pH 1
Color Yellow
CAS 67-63-0, 7732-18-5
Boiling Point 83°C
Packaging Glass Bottle
Specific Gravity 0.8
Quantity 100 mL
Molecular Formula HCl/C3H7OH
Physical Form Liquid
Synonym Hydrogen chloride solution
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.