Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 10% (v/v) in Methanol, Ricca Chemical
SDP

Catalog No. r3570200500
Click to view available options
Quantity:
500 mL

Chemical Identifiers

CAS 67-56-1, 7732-18-5, 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Appearance Clear colorless liquid
Odor Characteristic alcohol
Packaging Natural Poly Bottle
Assay Percent Range 10% (v/v)
Quantity 500 mL
Physical Form Solution
Solubility Information Solubility in Water: Infinite
Percent Purity 10%
Grade Laboratory
Chemical Name or Material Hydrochloric Acid in Methanol
Recommended Storage Flammable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.