Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrochloric Acid, 20° Be, Technical, Approx. 30%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
4 L
20 L
4 x 4 L
55 gal.
Packaging:
Drum
Poly Drum
Poly-Coated Glass Bottle
Description
Spectrum™ Chemical Hydrochloric Acid, 20 DEG Be, Technical is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids it is the least likely to be subject to an oxidation-reduction reaction. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis.
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly-Coated Glass Bottle |
| Quantity | 4 x 4 L |
| UN Number | UN1789 |
| Percent Purity | ∼30% |
| Grade | Technical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction