Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrochloric Acid, 3.00 N, Electronic Grade, Ricca Chemical

Click to view available options
Quantity:
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
Color | Colorless to Green-Yellow |
CAS Min % | 76.0, 22.19 |
CAS Max % | 79.0, 23.1 |
Packaging | Cubitainer |
Quantity | 10 L |
Physical Form | Liquid |
Grade | Electronic |
Concentration or Composition (by Analyte or Components) | 3.00 Normal |
Chemical Name or Material | Hydrochloric Acid |
Safety and Handling
ShelfLife | 18 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction