Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Hydrochloric Acid
Shop All Fisherbrand Products

Click to view available options
Quantity:
3 mL
Packaging:
8% v/v
Description
This product is an Educational Grade Chemical — no C of A is available.
Specifications
Specifications
Density | ∽1g/mL |
Packaging | 8% v/v |
Assay Percent Range | 3.65% v/v |
Quantity | 3 mL |
Physical Form | Liquid |
Linear Formula | HCl |
Formula Weight | 36.46 |
Chemical Name or Material | Hydrochloric Acid |
Safety and Handling
ShelfLife | 2 years |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction