Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrochloric Acid, 36.5 to 38.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Clear Glass Bottle
Clear PVC Coated Glass Bottle
Glass Bottle
PVC Coated Glass Bottle
Specifications
Specifications
| Packaging | PVC Coated Glass Bottle |
| Specific Gravity | 1L = 1.18kg |
| Quantity | 500 mL |
| IUPAC Name | hydrogen chloride |
| Formula Weight | 36.46 |
| Percent Purity | 36.5 to 38.0% |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Chemical Name or Material | Hydrochloric Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction