Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid, 6.0N Standardized Solution

Catalog No. p-4781933
Click to view available options
Quantity:
1 L
4 L
20 L

Catalog No. AA44921K2

Supplier: Thermo Scientific Chemicals 044921K2

Due to product restrictions, please to purchase or view availability for this product.

CAS: 7647-01-0 | ClH | 36.46 g/mol

6.0N HClsolution acts as a standard solution in volumetric titrations. In addition, it has industrial applications such as chemical transformations in hydrochlorination of rubber and production of vinyl and alkyl halide. In the semiconductor industry, it is used to etch semiconductor crystal and to purify silicon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0, 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless to Yellow
Solubility Miscible with water, methanol,ethanol and ether
Quantity 1 L
Formula Weight 36.46
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 49.6%; Water: 50.4%
Chemical Name or Material Hydrochloric acid
Hazard Category H290-H314-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8, Packing Group: II
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.