Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 6 M, Innovating Science™
SCI_ED

Catalog No. S031115
Click to view available options
Quantity:
500 mL
1 L

Chemical Identifiers

CAS 7732-18-5

Specifications

Concentration 6 M
Packaging HDPE
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Hydrochloric Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.