Learn More
Hydrochloric acid, 99.999999% (metals basis), 33% min
Description
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing. It finds use in the manufacture of polyvinyl chloride (PVC). Further, it is employed for regeneration of ion exchangers and in the tanning industry for the processing of leathers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
            Specifications
Specifications
| Color | Colorless to Yellow | 
| Boiling Point | 81.5°C to 110°C | 
| Concentration | ≤33% | 
| Density | 1.19 g/mL | 
| Odor | Pungent | 
| Quantity | 25 mL | 
| Physical Form | Liquid, Double Distilled In Quartz | 
| UN Number | UN1789 | 
| Merck Index | 14,4780 | 
| Solubility Information | Miscible with water. | 
| Show More | 
Safety and Handling
| Hazard Category | H290-H314-H335 | 
| Hazard Statement | GHS H Statement H314-H335 Causes severe skin burns and eye damage. May cause respiratory irritation.  | 
                                
| Precautionary Statement | P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c | 
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID | 
| EINECSNumber | 231-595-7 | 
| RTECSNumber | MW4025000 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.