Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrochloric acid concentrate, For 1 L standard solution, 1.0 M HCl (1.0 N), Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 100°C |
| Concentration | 1.0 M HCl (1.0 N) |
| CAS Max % | 96.5000%, 3.5000% |
| Density | 1.010 g/cm3 |
| Packaging | Ampoule |
| Vapor Pressure | 190 mmHg (25°C) |
| Quantity | 6 x 1 Ea. |
| Physical Form | Concentrate |
| Linear Formula | HCl |
| UN Number | UN1789 |
| Show More |
Safety and Handling
| Hazard Statement | P305 + P351 + P338 |
| ShelfLife | 1260 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Hydrochloric acid |
| Recommended Storage | Room Temp |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction