Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, ICP-OES, for Trace Metal Analysis

Catalog No. T003082500
Click to view available options
Quantity:
2.5 L

Suitable for digestion of samples prior to trace elemental analysis via ICP-OES. Tested for more than 20 metals at ppb levels and also meets ACS testing specifications. | CAS: 7647-01-0 | ClH | 36.46 g/mol

  • Suitable for trace metal analysis by ICP-OES
  • Tested for more than 20 metal impurities at ppb levels
  • Also meets general ACS testing specifications
  • Ideal for analysis of environmental samples
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color ≤ 5 (APHA)
Heavy Metals (as Pb) ≤ 1 ppm
Packaging PVC-Coated Glass Bottle
Residue after Ignition ≤ 5 ppm (ACS)
Quantity 2.5 L
Physical Form Liquid
Formula Weight 36.46
Percent Purity 36.5 to 38.0%
Grade ICP-OES
Chemical Name or Material Hydrochloric Acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.