Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrochloric Acid R1, 250 g/L HCl, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
Color | Colorless to Green-Yellow |
Concentration | 250g/L HCl |
Packaging | Natural Poly Bottle |
Quantity | 1 L |
Physical Form | Liquid |
Grade | Laboratory |
Chemical Name or Material | Hydrochloric Acid, Reagents |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction