Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid Solution, 1 N (Certified), Fisher Chemical PROMO

Catalog No. SA484FB25 Shop All Fisherbrand Products
Click to view available options
:
4 L

Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0, 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46 g/mol
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH 1.1
Melting Point 0°C
Boiling Point 100°C
Concentration 0.995 to 1.005 N
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84 L Potassium acid phthalate
Quantity 4 L
Physical Form Liquid
Grade Certified
Concentration or Composition (by Analyte or Components) 1 N
Chemical Name or Material Hydrochloric Acid Solution, 1 N
Show More Show Less
Hazard Category Signal Word: DANGER!
Hazard Statement CAUTION!
Precautionary Statement

Emergency Overview: Corrosive to metals. Irritating to eyes and skin. Harmful by inhalation, in contact with skin and if swallowed.

NFPA

Health:

DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.