Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hydrochloric Acid Solution, Sequencing Grade PROMO

Catalog No. PI24308PM
Click to view available options
:
Thermo Scientific™ Hydrochloric Acid Solution, Sequencing Grade

Constant-boiling 6N HCl solution, supplied in 1mL ampules, used to acid-hydrolyze proteins (protein hydrolysis) for HPLC amino acid composition analysis.

Highlights:

  • Hydrolyzes peptides in 6 hours at 150°C
  • Specially purified to give ninhydrin-negative blank on hydrolysis
  • Packaged in prescored ampules to eliminate contamination and ensure product integrity

Ordering Information:

X in the ordering table indicates that hazardous shipping charges apply.

Specifications

Product Type Hydrochloric Acid Solution
Content And Storage No product PDF available for storage information.
Quantity 10 x 1 mL
Form Solid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.