Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrochloric Acid, TraceSELECT™, 30.0 to 32.0 %, Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Plastic Bottle
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specifications
Specifications
| Color | Colorless |
| Melting Point | -35°C |
| Boiling Point | 42°C |
| Density | 1.15g/mL at 20°C, 1.2g/mL at 25°C |
| Packaging | Plastic Bottle |
| Vapor Pressure | 3.23psi (21.1°C), 7.93psi (37.7°C) |
| Quantity | 2.5 L |
| Physical Form | Liquid |
| UN Number | UN1789 |
| Beilstein | 1098214 |
| Show More |
Safety and Handling
| Hazard Statement | P261-P280-P305 + P351 + P338-P310 |
| Precautionary Statement | H290-H314-H335 |
| DOTInformation | 8/PGII |
| EINECSNumber | 231-595-7 |
| RTECSNumber | MW4025000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction