Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochlorothiazide, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603218
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

HY108, 58-93-5, C7H8ClN3O4S2

Spectrum™ Chemical Hydrochlorothiazide, USP is part of the thiazide class of diuretic drugs used for the treatment of congestive heart failure and hypertension. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-93-5
Molecular Formula C7H8ClN3O4S2
Molecular Weight (g/mol) 297.73
MDL Number MFCD00051765
InChI Key JZUFKLXOESDKRF-UHFFFAOYSA-N
IUPAC Name 6-chloro-1,1-dioxo-3,4-dihydro-2H-1λ⁶,2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)C1=CC2=C(NCNS2(=O)=O)C=C1Cl

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.