Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrocortisone, ≥97%, MP Biomedicals™

Catalog No. ICN10199610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10199610 10 g
ICN10199601 1 g
ICN10199605 5 g
ICN10199625 25 g
ICN10199680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10199610 Supplier MP Biomedicals, Inc Supplier No. 0210199610
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50-23-7
Molecular Formula C21H30O5
Molecular Weight (g/mol) 362.466
MDL Number MFCD00011654
InChI Key JYGXADMDTFJGBT-VWUMJDOOSA-N
Synonym hydrocortisone, cortisol, acticort, cetacort, cortef, 17-hydroxycorticosterone, hydrocortisyl, hydrasson, cobadex, cortril
PubChem CID 5754
ChEBI CHEBI:17650
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4(C(=O)CO)O)C)O

Specifications

Color White
Assay Percent Range ≥97%
Quantity 10 g
Formula Weight 362.5
Physical Form Crystalline Powder
Chemical Name or Material Hydrocortisone
EINECSNumber 200-020-1
Recommended Storage Store at Room Temperature (15 to 30°C), protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.