Learn More
Thermo Scientific Chemicals Hydrocortisone, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Hydrocortisone is the synthetic formulation of the naturally occurring hormone, cortisol
• It is available as a white crystalline powder soluble in water, DMSO, and ethanol
• It is stable at room temperature but sensitive to light, and reactive in the presence of strong oxidizing agents
Applications
• Hydrocortisone works as an agonist to glucocorticoid receptors, thereby suppressing the inflammatory response of the immune system
• It’s administration induces gluconeogenesis, thus increasing endogenous blood sugar levels
• Research indicates a role for hydrocortisone in the metabolism of fat, protein, and carbohydrates
Specifications
Specifications
| Melting Point | 212°C to 217°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 25 g |
| Merck Index | 15, 4824 |
| Specific Rotation Condition | +150° (156°C c=1,dioxane) |
| Specific Rotation | + 150 |
| Solubility Information | Solubility in water: 0.28g/L (25°C). Other solubilities: soluble in sulfuric acid,partially soluble in alcohol and propylene glycol,soluble in dioxane; sol (g/l) (25°C): chcl3 1.6;,ether 0.35; etoh 15; meoh 6.2; acetone 9.3;,propylene glycol 1 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May damage the unborn child. Suspected of damaging fertility. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. |
| EINECSNumber | 200-020-1 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.