Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrocortisone Acetate, Micronized, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18603222
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle

CO138, 50-03-3, C23H32O6

Spectrum™ Chemical Hydrocortisone Acetate, Micronized, USP is a topical corticosteroid product used to help treat eczema, psoriasis and other skin conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-03-3
Molecular Formula C23H32O6
Molecular Weight (g/mol) 404.50
MDL Number MFCD00037714
InChI Key ALEXXDVDDISNDU-JZYPGELDSA-N
IUPAC Name 2-[(1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation +158° to +167°
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Hydrocortisone Acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.