Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrocortisone, Micronized, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18603234 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-234 25 g Amber Glass Bottle
18-603-233 5 g Amber Glass Bottle
18-603-235 100 g Amber Glass Bottle
18-603-238 500 g Amber Glass Bottle
18-603-236 1 kg Amber Glass Bottle
18-603-237 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-234 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CO13725GM
Add to Cart
Add to Cart

CO137, 50-23-7, C21H30O5

Spectrum™ Chemical Hydrocortisone, Micronized, USP , otherwise known as Cortisol, is a glucocorticoid, a steroid hormone. Secreted in the adrenal cortex its function is to increase blood sugar through a process called gluconeogenesis. It suppresses the immune system and helps in the metabolism of protein, fat and carbohydrate. Recreated synthetically, it has been used to treat a variety of diseases. It can be administered by injection or orally and is used as an immunosuppressant to help treat severe allergic reactions. It can be administered locally and topically to help treat allergies such as eczema, psoriasis and other skin conditions. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-23-7
Molecular Formula C21H30O5
Molecular Weight (g/mol) 362.47
InChI Key JYGXADMDTFJGBT-VWUMJDOOSA-N
IUPAC Name (1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Loss on Drying 1%
Melting Range 217°C to 220°C
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation +150° to +156°
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Hydrocortisone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.