Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric Acid, 0.1% (w/w), Ricca Chemical
SDP

Catalog No. r38153004a Shop All Ricca Chemical Company Products
Click to view available options
:
Package: Natural Poly; Size: 4 L

Chemical Identifiers

CAS 7732-18-5, 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Appearance of Solution Colorless Liquid
Concentration 0.095 to 0.105 N
CAS Max % 0.1, 99.9
Packaging Natural Poly Bottle
Quantity 4 L
Percent Purity 0.1% W/W
Grade Laboratory
Chemical Name or Material Hydrofluoric Acid
ShelfLife 24 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.