Learn More
Sulfuric Acid 96% LM Prime, Transene

Description
Designed for electronics manufacturing are referred to as Electronic Grade solvents. These solvents contain very low levels of heavy metals, typically measured in parts per billion or parts per trillion. They are commonly utilized as cleaners and etchants in the production of semiconductors and microprocessors. 96% Sulfuric Acid Electronic/Cleanroom Grade is suitable for use in electronics and related fields.
- Industries:
- Electronics and Semiconductor
- Cleaning agents
Safety and Shipping Information: Sulfuric Acid from Lab Alley is classified as hazardous, and a hazmat fee is required. DOT: Sulfuric Acid, 8, UN1830, PG II

Specifications
Specifications
Packaging | HDPE |
Quantity | 55 Gallon |
Percent Purity | 96% |
Grade | Electronic |
Chemical Name or Material | Sulfuric Acid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.