Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrofluoric Acid, ACS, 48-51%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 mL
1 gal.
6 x 500 mL
4 x 1 gal.
200 L
Packaging:
Poly Bottle
Description
Spectrum™ Chemical Hydrofluoric Acid, 48 Percent, Suitable for Electronic Use, Reagent, ACS is a highly corrosive solution of hydrogen fluoride in water is capable of dissolving a wide range of materials including oxides and glass. Used in a variety of fields including oil refining and production of fluorides it can also be used to dissolve rock samples before analysis. As an ACS grade Reagent, Spectrum Chemical manufactured hydrofluoric acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications
Specifications
CAS Max % | 100% |
Heavy Metals (as Pb) | 0.5 ppm |
Packaging | Poly Bottle |
Residue after Ignition | 5 ppm |
Quantity | 200 L |
UN Number | UN1790 |
Percent Purity | 48 to 51% |
Grade | ACS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction