Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric Acid, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Bagged Poly Bottle
Narrow Mouth Poly Bottle

Chemical Identifiers

7664-39-3
FH
20.006
KRHYYFGTRYWZRS-UHFFFAOYSA-N
14917
CHEBI:29228

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use
Quantity 500mL
SMILES F
IUPAC Name fluorane
Formula Weight 20.01
Packaging Narrow Mouth Poly Bottle
Specific Gravity 1L = 1.18kg
Chemical Name or Material Hydrofluoric Acid
Grade BAKER INSTRA-ANALYZED™ Plus