Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen chloride, 1M soln. in ethyl acetate

Catalog No. p-7031257
Click to view available options
Quantity:
100 mL
500 mL

CAS: 7647-01-0 | ClH | 36.46 g/mol

In chemical industry production of inorganic salts, Production of vinyl chloride from acetylene, alkyl chlorides from olefins, dyes, artificial silk, pigment, pharmaceutical hydrochloride, As catalyst and solvent in organic syntheses, Alcohol chlorination reagent, Nitration reaction, Manufacture of phosphoric acid and in the production of ammonium chloride, Neutralization of basic systems, Cleaning agents. Hydrolyzing of starch and proteins in the preparation of various food products. The hydrogen chloride gas is mainly used for EDC and vinyl chloride monomer (VCM) production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Linear Formula HCl
UN Number UN2924
Concentration 1M soln. in ethyl acetate
Density 0.9
Solubility Information Reacts with water.
IUPAC Name hydrogen chloride
Formula Weight 36.46
Quantity 100 mL
Chemical Name or Material Hydrochloric acid
Hazard Category H225-H315-H318-H335-H336
Hazard Statement GHS H Statement
H225-H318-H332-H336-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if inhaled.
May cause drowsiness or dizziness.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.