Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hydrogen exchanger 3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988673
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8673 Supplier Bioss Supplier No. BS8601R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NHE3 (also known as SLC9A3 - Sodium/hydrogen exchanger 3) is a member of the sodium/hydrogen exchanger family that plays a crucial role in acid-base and volume homeostasis by mediating the majority of sodium and bicarbonate reabsorption in the proximal tubule of the kidney. Mutations in the gene can lead to Diarrhea 8.

Specifications

Antigen hydrogen exchanger 3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC9A3
Gene Accession No. P48764
Gene Alias 9030624O13Rik; AI930210; BOS_19957; DIAR8; MGC126718; MGC126720; Na(+)/H(+) exchanger 3; Na/H exchanger 3; Na+/H+ exchanger homolog; NHE3; NHE-3; NHE-3 antibody; plasma membrane ion transport protein; putative sodium hydrogen exchanger subtype 3; Slc9a3; SLC9A3 antibody; sodium proton exchanger; Sodium/hydrogen exchanger 3; Solute carrier family 9 (sodium/hydrogen exchanger 3), antiporter 3, Na+/H+ (amiloride insensitive); solute carrier family 9 (sodium/hydrogen exchanger), isoform 3; solute carrier family 9 (sodium/hydrogen exchanger), member 3; Solute carrier family 9 member 3; solute carrier family 9 member A3; solute carrier family 9, member 3; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3
Gene Symbols SLC9A3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NHE3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6550
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.