Learn More
Hydrogen hexafluoroantimonate(V), ≈65% aq. soln., Thermo Scientific Chemicals
Description
Hydrogen hexafluoroantimonate(V) is an excellent medium for the preparation of alkylidene oxonium salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Orange |
| Linear Formula | HF·SbF5 |
| Quantity | 25 g |
| UN Number | UN3264 |
| Solubility Information | It is fully miscible with water,soluble in SO2ClF,SO2. |
| Formula Weight | 236.75 |
| Concentration or Composition (by Analyte or Components) | ≈65% aq. soln. |
| Physical Form | Liquid |
| Chemical Name or Material | Hydrogen hexafluoroantimonate(V) |
Safety and Handling
| Hazard Category | H302+H332-H314-H335 |
| Hazard Statement | GHS H Statement H314-H302-H332 Causes severe skin burns and eye damage. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 241-023-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.