Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 3% Solution, Reagent, 3.0-3.5%, Spectrum™

Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner H1060200LTBL

 View more versions of this product

Catalog No. 18-610-947


Due to product restrictions, please to purchase or view availability for this product.


Specifications

Specifications

7722-84-1, 7732-18-5
To Pass Test
3.0 to 3.5%
5e-06
5e-05
3e-05
1.01
200 L
5e-07
1e-05
Chemical Solution
5e-06
Poly Drum
0.0002
0.0001
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.