Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 3% (w/w), Stabilized Reagent Grade, Ricca Chemical
SDP

Catalog No. 381955 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
55 gal.
Packaging:
Amber Poly Bottle
Black Poly Bottle
Hard Pack
Poly Drum

Chemical Identifiers

CAS 7722-84-1, 7732-18-5, 103-84-4
Molecular Formula H2O2
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name peroxol
SMILES OO

Specifications

Color Colorless
CAS Min % 2.0, 96.0, 0.02
CAS Max % 4.0, 98.0, 0.02
Molecular Weight (g/mol) 34.01
Percent Purity 3%
Grade Reagent
Packaging Poly Drum
Treatment(s) Stabilized
Solvent or Matrix Aqueous Solution
Quantity 55 gal.
Physical Form Liquid
Concentration or Composition Notes 3% (w/w)
Chemical Name or Material Hydrogen Peroxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.