Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 30%, Solution, Reagent, 29.0-32.0%, Spectrum™ Chemical
SDP

Catalog No. p-7165642 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mL
500 mL
4 L
200 L
Packaging:
Drum
Poly Bottle

Specifications

Chemical Name or Material Ferrous Ammonium Sulfate
Color light green
CAS 7783-85-9, 7732-18-5, 7664-93-9
Concentration 0.1 N ± 0.006 N
Percent Purity 29 to 32%
Grade Reagent
Packaging Drum
Residue after Evaporation 0.002%
Specific Gravity 1.1
Quantity 200 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.