Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 30% (w/w), ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. 3821732
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Vented Natural Poly Bottle
Vented Poly Bottle

Chemical Identifiers

CAS 7722-84-1, 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
CAS Min % 25.0, 65.0
CAS Max % 35.0, 75.0
Percent Purity 30%
Grade ACS Reagent
Concentration or Composition (by Analyte or Components) 30% (w/w)
Packaging Vented Poly Bottle
pH 3.3
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.