Learn More
Hydrogen Peroxide, 35% Solution, FCC, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Hydrogen Peroxide, 35 Percent Solution, FCC is used as a disinfectant in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
Acidity | 0.03% |
CAS Max % | 65%, 35% |
UN Number | UN2014 |
Concentration | 35% |
Percent Purity | 35% |
Grade | FCC |
Packaging | Poly Bottle |
Quantity | 500 mL |
Chemical Name or Material | Hydrogen Peroxide |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.