Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen peroxide Solution, 25-35%, ≥30% (RT), MilliporeSigma™ Supelco™

Catalog No. 111017944 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7944 5 L
11-101-8036 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-7944 Supplier MilliporeSigma Supelco Supplier No. 169115LF
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00011333

Specifications

Density 1.11 g/mL (at 20°C)
CAS 7722-84-1
Linear Formula H2O2
Quantity 5 L
Vapor Pressure 23.3 mmHg (30°C)
Grade For ultratrace analysis
Concentration or Composition (by Analyte or Components) 25-35%
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.