Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide Solution, ACS, 29-32%, Spectrum™ Chemical
SDP

Catalog No. 18603251 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 100 mL
6 x 500 mL
4 x 4 L
200 L
Packaging:
Poly Bottle

H1065, 7722-84-1, H2O2

Spectrum™ Chemical Hydrogen Peroxide Solution, 30 Percent, Reagent, ACS, is the simplest peroxide, a strong oxidizer that appears as a clear and slightly viscous liquid. Its oxidizing properties are so strong that it is considered a highly reactive oxygen species. Due to it being a weak acid it can form hydroperoxide or peroxide salts. One of its biggest uses is in the treatment of drinking water, removing odors, organic materials and the amount of Hydrogen sulfides and Iron. As an ACS grade Reagent, Spectrum Chemical manufactured Hydrogen Peroxide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Specifications

Chemical Name or Material Hydrogen Peroxide Solution
Color Colorless
CAS 7722-84-1, 7732-18-5
CAS Max % 30%, 70%
UN Number UN2014
Concentration 30%
Percent Purity 29 to 32%
Grade ACS
Packaging Poly Bottle
Residue after Evaporation 0.002%
Quantity 6 x 100 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.